ENAH90821F85
Enamine
lithium(1+) 3-(3-hydroxypropoxy)propanoate
About This Item
Iupac Name
lithium 3-(3-hydroxypropoxy)propanoate
Inchi Code
1S/C6H12O4.Li/c7-3-1-4-10-5-2-6(8)9;/h7H,1-5H2,(H,8,9);/q;+1/p-1
InChI key
HFZMXSYURIKBGY-UHFFFAOYSA-M
Salt Data
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-7562783
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service