Skip to Content
Merck
All Photos(1)

Key Documents

ATE423888789

AstaTech, Inc.

3-AMINO-2,2-DIMETHYL-1-PROPANOL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
103.16
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

~ 70 C

Iupac Name

3-amino-2,2-dimethyl-1-propanol

Inchi Code

1S/C5H13NO/c1-5(2,3-6)4-7/h7H,3-4,6H2,1-2H3

InChI key

FNVOFDGAASRDQY-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=35559

Melting Point

~ 70 C

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=35391


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service