Skip to Content
Merck
All Photos(1)

Key Documents

APO455834906

Apollo Scientific Ltd

4-Chloro-2-fluorobenzylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€59.80
5 G
€66.40
25 G
€153.30

Vendor SKU: PC1864FD-1G

€59.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
€59.80
5 G
€66.40
25 G
€153.30

About This Item

CAS Number:
Molecular Weight:
196.05
MDL number:

Vendor SKU: PC1864FD-1G

€59.80


Ships in 14 days - from Aldrich Partner.

bp

243-245

Iupac Name

(4-chloro-2-fluorophenyl)methanamine hydrochloride

Inchi Code

1S/C7H7ClFN.ClH/c8-6-2-1-5(4-10)7(9)3-6;/h1-3H,4,10H2;1H

InChI key

DFRJZBWKVAXYRV-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-chloro-2-fluorobenzylamine-hydrochloride

Synonym

(4-Chloro-2-fluorophenyl)methylamine hydrochloride

Boiling point

243-245

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-chloro-2-fluorobenzylamine-hydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service