Skip to Content
Merck
All Photos(1)

Documents

W201006

Sigma-Aldrich

trans-Aconitic acid

≥98%, FCC

Synonym(s):

trans-Propene-1,2,3-tricarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C(CO2H)=CHCO2H
CAS Number:
Molecular Weight:
174.11
FEMA Number:
2010
Beilstein:
1725830
EC Number:
Council of Europe no.:
33
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

reg. compliance

FCC

Assay

≥98%

mp

190 °C (dec.) (lit.)

application(s)

flavors and fragrances

Organoleptic

cashew; meaty; musty; vegetable

SMILES string

OC(=O)C\C(=C/C(O)=O)C(O)=O

InChI

1S/C6H6O6/c7-4(8)1-3(6(11)12)2-5(9)10/h1H,2H2,(H,7,8)(H,9,10)(H,11,12)/b3-1+

InChI key

GTZCVFVGUGFEME-HNQUOIGGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes.
Taste at 25 ppm

Packaging

Packaged in glass bottles

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayin Pang et al.
Physiologia plantarum, 154(4), 511-525 (2014-10-08)
The aim of this study was to investigate the capacity of three perennial legume species to access sources of varyingly soluble phosphorus (P) and their associated morphological and physiological adaptations. Two Australian native legumes with pasture potential (Cullen australasicum and
K Tanaka et al.
Journal of chromatography, 239, 301-322 (1982-04-30)
Gas chromatographic retention indices have been compiled for 163 metabolically important compounds (mostly organic acids) in the form of methylene units, as trimethylsilyl derivatives, on 10% OV-1 and 10% OV-17 columns. Comprehensive references on metabolic diseases that can be diagnosed
Mosciano, G.
Perfumer & Flavorist, 21, 51-51 (1996)
M Y Tsai et al.
Journal of chromatography, 341(1), 1-10 (1985-05-31)
To offer a procedure with increased resolution compared to packed-column gas chromatography, we developed a dual-capillary method of gas chromatography for diagnosing organic acidurias. We derivatized and injected organic acids repeatedly on two different bonded phase columns (DB-1, DB-1701) to
[Uremic peak 2a in high performance liquid chromatography--acidic components and their membrane permeability].
K Koide et al.
Nihon Jinzo Gakkai shi, 28(11), 1481-1489 (1986-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service