Skip to Content
Merck
All Photos(1)

Key Documents

653500

Sigma-Aldrich

Ethyl 2-amino-4-(4-bromophenyl)thiazole-5-carboxylate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11BrN2O2S
CAS Number:
Molecular Weight:
327.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

204-208 °C

SMILES string

CCOC(=O)c1sc(N)nc1-c2ccc(Br)cc2

InChI

1S/C12H11BrN2O2S/c1-2-17-11(16)10-9(15-12(14)18-10)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H2,14,15)

InChI key

HZNNHEOEYQXAAE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service