Skip to Content
Merck
All Photos(1)

Documents

571903

Sigma-Aldrich

Diethyl 1-benzyl-3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO6
CAS Number:
Molecular Weight:
333.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

143-146 °C (lit.)

SMILES string

CCOC(=O)c1c(O)c(O)c(C(=O)OCC)n1Cc2ccccc2

InChI

1S/C17H19NO6/c1-3-23-16(21)12-14(19)15(20)13(17(22)24-4-2)18(12)10-11-8-6-5-7-9-11/h5-9,19-20H,3-4,10H2,1-2H3

InChI key

AXYCCUUFBQELMR-UHFFFAOYSA-N

Application

Starting material for substituted 3,4-ethylenedioxypyrroles.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zong, K.; Abboud, K.; Reynolds, J.
Tetrahedron Letters, 45, 4973-4973 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service