Skip to Content
Merck
All Photos(1)

Documents

470465

Sigma-Aldrich

tert-Dodecyl polysulfide

average Mn ~434, TPS® 20

Synonym(s):

Di-tert-dodecyl polysulfide, TPS® 20

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H19C(CH3)2)2Sx
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

liquid

mol wt

average Mn ~434

manufacturer/tradename

TPS® 20

refractive index

n20/D 1.511

viscosity

53 cP(40 °C)(lit.)

transition temp

pour point −21 °C

density

0.95 g/mL at 25 °C

SMILES string

CCCCCCCCCC(C)(C)SSC(C)(C)CCCCCCCCC

InChI

1S/C24H50S3/c1-17(19(3,4)5)21(9,10)23(13,14)25-27-26-24(15,16)22(11,12)18(2)20(6,7)8/h17-18H,1-16H3

InChI key

AWSNFBCZTJENAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Arkema Inc.
TPS is a registered trademark of Arkema France
The Gene Construction Kit is a trademark of Textco

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service