Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458031

Sigma-Aldrich

Neopentyl glycol dibenzoate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
(C6H5CO2CH2)2C(CH3)2
CAS Number:
Molecular Weight:
312.36
EC Number:
MDL number:
UNSPSC Code:
12352300

mp

45-51 °C (lit.)

SMILES string

CC(C)(COC(=O)c1ccccc1)COC(=O)c2ccccc2

Application

Plasticizer

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service