Skip to Content
Merck
All Photos(1)

Key Documents

425702

Sigma-Aldrich

Praseodymium(III) trifluoromethanesulfonate

98%

Synonym(s):

Praseodymium(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Pr
CAS Number:
Molecular Weight:
588.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: praseodymium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

SMILES string

[Pr+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Pr/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

ROUBZIWQWFQCHU-UHFFFAOYSA-K

General description

Praseodymium (III) trifluoromethanesulfonate (Praseodymium(III) triflate) is a water-tolerant Lewis acid. It is an efficient catalyst employed for the synthesis of α-aminonitrile, via condensation of aldehydes, amines and trimethylsilyl cyanide.

Application

Catalyst for:
  • Asymmetric α-amination reactions
  • Polymerization reactions
  • Regioselective reductive ring opening reactions
  • Intermolecular hydroamination reactions
  • Paal-Knorr condensation
  • Friedel-Crafts alkenylation
  • Friedel-Crafts acylation
  • Biginelli reactions
Praseodymium(III) trifluoromethanesulfonate (Praseodymium(III) triflate) was employed as catalyst for the aqueous aza-Diels-Alder reactions of aldehydes and dienes. It was also used as catalyst for the Aldol reaction of silyl enol ethers with aldehydes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu L, Chen D, Wang PG
Tetrahedron Letters, 37, 2169-2169 (1996)
Praseodymium Trifluoromethylsulfonate as an Efficient and Recyclable Catalyst for the Synthesis of a-Aminonitriles.
De SK and Gibbs RA.
Synthetic Communications, 35(7), 961-966 (2005)
The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service