Skip to Content
Merck
All Photos(2)

Documents

110221

Sigma-Aldrich

Acrolein

contains hydroquinone as stabilizer, 90%

Synonym(s):

2-Propenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCHO
CAS Number:
Molecular Weight:
56.06
Beilstein:
741856
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

1.94 (vs air)

vapor pressure

4.05 psi ( 20 °C)

Assay

90%

form

liquid

autoignition temp.

428 °F

contains

hydroquinone as stabilizer

expl. lim.

31 %

impurities

<10% water and cyclic dimer

refractive index

n20/D 1.403 (lit.)

bp

53 °C (lit.)

mp

−87 °C (lit.)

solubility

H2O: soluble 2
alcohol: soluble
benzene: miscible
diethyl ether: miscible

density

0.839 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)C=C

InChI

1S/C3H4O/c1-2-3-4/h2-3H,1H2

InChI key

HGINCPLSRVDWNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acrolein causes transient urothelial loss and exposes local afferent terminals to a toxic environment.
Acrolein is a metabolite that is generated by catabolic pathaways via many oxidases.

Application

Acrolein has been used in the preparation of monomeric and dimeric β-hydroxypropionaldehyde in a hydration reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Talarico et al.
Antimicrobial agents and chemotherapy, 33(5), 674-679 (1989-05-01)
Lactobacillus reuteri converts glycerol into a potent cell growth inhibitor. This substance, termed reuterin, inhibits the growth of gram-positive and gram-negative bacteria as well as yeasts, fungi, and protozoa. Semipreparative chromatography was used to purify reuterin, and Fourier transform infrared
Anthony E Pegg
Chemical research in toxicology, 26(12), 1782-1800 (2013-11-15)
Polyamines are ubiquitous and essential components of mammalian cells. They have multiple functions including critical roles in nucleic acid and protein synthesis, gene expression, protein function, protection from oxidative damage, the regulation of ion channels, and maintenance of the structure
Giancarlo Aldini et al.
Molecular nutrition & food research, 55(9), 1301-1319 (2011-08-02)
Acrolein (ACR) is a toxic and highly reactive α,β-unsaturated aldehyde widely distributed in the environment as a common pollutant and generated endogenously mainly by lipoxidation reactions. Its biological effects are due to its ability to react with the nucleophilic sites
Riyi Shi et al.
Molecular nutrition & food research, 55(9), 1320-1331 (2011-08-09)
Acrolein, an α,β-unsaturated aldehyde, is a ubiquitous pollutant that is also produced endogenously through lipid peroxidation. This compound is hundreds of times more reactive than other aldehydes such as 4-hydroxynonenal, is produced at much higher concentrations, and persists in solution
S Vamsee Raju et al.
American journal of respiratory and critical care medicine, 188(11), 1321-1330 (2013-09-18)
Several extrapulmonary disorders have been linked to cigarette smoking. Smoking is reported to cause cystic fibrosis transmembrane conductance regulator (CFTR) dysfunction in the airway, and is also associated with pancreatitis, male infertility, and cachexia, features characteristic of cystic fibrosis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service