Merck
GC Analysis of Durability Test of Molded Thermogreen® LB-2 Septum on SLB-5ms
GC Analysis of Durability Test of Molded Thermogreen® LB-2 Septum on SLB-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
related product
Product No.
Description
Pricing
Nonpolar Column Test Mix
certified reference material, 500 μg/mL in dichloromethane
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
115 °C (15 min)
inj. temp.
250 °C
detector
FID, 360 °C
carrier gas
helium, 30 cm/sec. @ 115 °C
injection
1 μL, 100:1 split
liner
4 mm I.D., split, cup design, unpacked
sample
Nonpolar Column Test Mix, each analyte at 500 μg/mL in methylene chloride (47300-U)
Description
Legal Information
Thermogreen is a registered trademark of Merck KGaA, Darmstadt, Germany