GC Analysis of 2-Hydroxy-2-Methyl Butyric Acid Methyl Ester Enantiomers on Astec® CHIRALDEX™ B-DP
CONDITIONS
column
Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 μm (78023AST)
oven
70 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: 2-hydroxy-2-methyl butyric acid methyl ester enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC