Merck
  • Separation characteristics of fatty acid methyl esters using SLB-IL111, a new ionic liquid coated capillary gas chromatographic column.
Separation characteristics of fatty acid methyl esters using SLB-IL111, a new ionic liquid coated capillary gas chromatographic column.
Journal of chromatography. A (2010-12-24)
Pierluigi Delmonte, Ali-Reza Fardin Kia, John K G Kramer, Magdi M Mossoba, Len Sidisky, Jeanne I Rader
ABSTRACT
The ionic liquid SLB-IL111 column, available from Supelco Inc., is a novel fused capillary gas chromatography (GC) column capable of providing enhanced separations of fatty acid methyl esters (FAMEs) compared to the highly polar cyanopropyl siloxane columns currently recommended for the separation of cis- and trans isomers of fatty acids (FAs), and marketed as SP-2560 and CP-Sil 88. The SLB-IL111 column was operated isothermal at 168°C, with hydrogen as carrier gas at 1.0 mL/min, and the elution profile was characterized using authentic GC standards and synthetic mono-unsaturated fatty acids (MUFAs) and conjugated linoleic acid (CLA) isomers as test mixtures. The SLB-IL111 column provided an improved separation of cis- and trans-18:1 and cis/trans CLA isomers. This is the first direct GC separation of c9,t11- from t7,c9-CLA, and t15-18:1 from c9-18:1, both of which previously required complimentary techniques for their analysis using cyanopropyl siloxane columns. The SLB-IL111 column also provided partial resolution of t13/t14-18:1, c8- from c6/c7-18:1, and for several t,t-CLA isomer pairs. This column also provided elution profiles of the geometric and positional isomers of the 16:1, 20:1 and 18:3 FAMEs that were complementary to those obtained using the cyanopropyl siloxane columns. However, on the SLB-IL111 column the saturated FAs eluted between the cis- and trans MUFAs unlike cyanopropyl siloxane columns that gave a clear separation of most saturated FAs. These differences in elution pattern can be exploited to obtain a more complete analysis of complex lipid mixtures present in ruminant fats.
Product Description
Supelco
SP®-2560 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.20 μm
Pricing and availability is not currently available.
Supelco
SP®-2560 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.20 μm
Pricing and availability is not currently available.
Supelco
SP®-2560 Capillary GC Column, L × I.D. 75 m × 0.18 mm, df 0.14 μm
Pricing and availability is not currently available.
Supelco
SP®-2560 Capillary GC Column, L × I.D. 200 m × 0.25 mm, df 0.20 μm
Pricing and availability is not currently available.