Merck
  • Toxicological significance of renal Bcrp: Another potential transporter in the elimination of mercuric ions from proximal tubular cells.
Toxicological significance of renal Bcrp: Another potential transporter in the elimination of mercuric ions from proximal tubular cells.
Toxicology and applied pharmacology (2015-04-15)
Christy C Bridges, Rudolfs K Zalups, Lucy Joshee
ABSTRACT
Secretion of inorganic mercury (Hg(2+)) from proximal tubular cells into the tubular lumen has been shown to involve the multidrug resistance-associated protein 2 (Mrp2). Considering similarities in localization and substrate specificity between Mrp2 and the breast cancer resistance protein (Bcrp), we hypothesize that Bcrp may also play a role in the proximal tubular secretion of mercuric species. In order to test this hypothesis, the uptake of Hg(2+) was examined initially using inside-out membrane vesicles containing Bcrp. The results of these studies suggest that Bcrp may be capable of transporting certain conjugates of Hg(2+). To further characterize the role of Bcrp in the handling of mercuric ions and in the induction of Hg(2+)-induced nephropathy, Sprague-Dawley and Bcrp knockout (bcrp(-/-)) rats were exposed intravenously to a non-nephrotoxic (0.5 μmol · kg(-1)), a moderately nephrotoxic (1.5 μmol · kg(-1)) or a significantly nephrotoxic (2.0 μmol · kg(-1)) dose of HgCl2. In general, the accumulation of Hg(2+) was greater in organs of bcrp(-/-) rats than in Sprague-Dawley rats, suggesting that Bcrp may play a role in the export of Hg(2+) from target cells. Within the kidney, cellular injury and necrosis was more severe in bcrp(-/-) rats than in controls. The pattern of necrosis, which was localized in the inner cortex and the outer stripe of the outer medulla, was significantly different from that observed in Mrp2-deficient animals. These findings suggest that Bcrp may be involved in the cellular export of select mercuric species and that its role in this export may differ from that of Mrp2.
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Sodium 2,3-dimercaptopropanesulfonate monohydrate, 95%