Merck
  • First dose in neonates: are juvenile mice, adults and in vitro-in silico data predictive of neonatal pharmacokinetics of fluconazole.
First dose in neonates: are juvenile mice, adults and in vitro-in silico data predictive of neonatal pharmacokinetics of fluconazole.
Clinical pharmacokinetics (2014-08-27)
Wei Zhao, Chantal Le Guellec, Daniel K Benjamin, William W Hope, Thomas Bourgeois, Kevin M Watt, Johannes N van den Anker, Boris Matrot, Harri Saxen, Kalle Hoppu, Paolo Manzoni, Evelyne Jacqz-Aigrain
ABSTRACT
Selection of the first-dose-in-neonates is challenging. The objective of this proof-of-concept study was to evaluate a pharmacokinetic bridging approach to predict a neonatal dosing regimen. We selected fluconazole as a paradigm compound. We used data from studies in juvenile mice and adults to develop population pharmacokinetic models using NONMEM. We also develop a physiologically-based pharmacokinetic model from in vitro-in silico data using Simcyp. These three models were then used to predict neonatal pharmacokinetics and dosing regimens for fluconazole. From juvenile mice to neonates, a correction factor of maximum lifespan potential should be used for extrapolation, while a "renal factor" taking into account renal maturation was required for successful bridging based on adult and in vitro-in silico data. Simulations results demonstrated that the predicted drug exposure based on bridging approach was comparable to the observed value in neonates. The prediction errors were -2.2, +10.1 and -4.6 % for juvenile mice, adults and in vitro-in silico data, respectively. A model-based bridging approach provided consistent predictions of fluconazole pharmacokinetic parameters in neonates and demonstrated the feasibility of this approach to justify the first-dose-in-neonates, based on all data available from different sources (including physiological informations, preclinical studies and adult data), allowing evidence-based decisions of neonatal dose rather than empiricism.
Product Description
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard