Merck
  • Early reperfusion and clinical outcomes in patients with M2 occlusion: pooled analysis of the PROACT II, IMS, and IMS II studies.
Early reperfusion and clinical outcomes in patients with M2 occlusion: pooled analysis of the PROACT II, IMS, and IMS II studies.
Journal of neurosurgery (2014-09-27)
Ralph Rahme, Sharon D Yeatts, Todd A Abruzzo, Lincoln Jimenez, Liqiong Fan, Thomas A Tomsick, Andrew J Ringer, Anthony J Furlan, Joseph P Broderick, Pooja Khatri
ABSTRACT
The role of endovascular therapy in patients with acute ischemic stroke and a solitary M2 occlusion remains unclear. Through a pooled analysis of 3 interventional stroke trials, the authors sought to analyze the impact of successful early reperfusion of M2 occlusions on patient outcome. Patients with a solitary M2 occlusion were identified from the Prolyse in Acute Cerebral Thromboembolism (PROACT) II, Interventional Management of Stroke (IMS), and IMS II trial databases and were divided into 2 groups: successful reperfusion (thrombolysis in cerebral infarction [TICI] 2-3) at 2 hours and failed reperfusion (TICI 0-1) at 2 hours. Baseline characteristics and clinical outcomes were compared. Sixty-three patients, 40 from PROACT II and 23 from IMS and IMS II, were identified. Successful early angiographic reperfusion (TICI 2-3) was observed in 31 patients (49.2%). No statistically significant difference in the rates of intracerebral hemorrhage (60.9% vs 47.6%, p = 0.55) or mortality (19.4% vs 15.6%, p = 0.75) was observed. However, there was a trend toward higher incidence of symptomatic hemorrhage in the TICI 2-3 group (17.4% vs 0%, p = 0.11). There was also a trend toward higher baseline glucose levels in this group (151.5 mg/dl vs 129.6 mg/ dl, p = 0.09). Despite these differences, the rate of functional independence (modified Rankin Scale Score 0-2) at 3 months was similar (TICI 2-3, 58.1% vs TICI 0-1, 53.1%; p = 0.80). A positive correlation between successful early reperfusion and clinical outcome could not be demonstrated for patients with M2 occlusion. Irrespective of reperfusion status, such patients have better outcomes than those with more proximal occlusions, with more than 50% achieving functional independence at 3 months.
Sigma-Aldrich
6-Maleimidohexanoic acid, ≥98.0% (HPLC)
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)