Merck
  • Participation of glutamate-333 of the D1 polypeptide in the ligation of the Mn₄CaO₅ cluster in photosystem II.
Participation of glutamate-333 of the D1 polypeptide in the ligation of the Mn₄CaO₅ cluster in photosystem II.
Biochemistry (2013-10-31)
Rachel J Service, Junko Yano, Preston L Dilbeck, Robert L Burnap, Warwick Hillier, Richard J Debus
ABSTRACT
In the 1.9 Å structural model of photosystem II (PDB: 3ARC), the amino acid residue Glu333 of the D1 polypeptide coordinates to the oxygen-evolving Mn₄CaO₅ cluster. This residue appears to be highly significant in that it bridges the two Mn ions (Mn(B3) and the "dangling" Mn(A4)) that are also bridged by the oxygen atom O5. This oxygen atom has been proposed to be derived from one of two substrate water molecules and to become incorporated into the product dioxygen molecule during the final step in the catalytic cycle. In addition, the backbone nitrogen of D1-Glu333 interacts directly with a nearby Cl⁻ atom. To further explore the influence of this structurally unique residue on the properties of the Mn₄CaO₅ cluster, the D1-E333Q mutant of the cyanobacterium Synechocystis sp. PCC 6803 was characterized with a variety of biophysical and spectroscopic methods, including polarography, EPR, X-ray absorption, and FTIR difference spectroscopy. The kinetics of oxygen release in the mutant were essentially unchanged from those in wild-type. In addition, the oxygen flash yields exhibited normal period-four oscillations having normal S state parameters, although the yields were lower, indicative of the mutant's lower steady-state dioxygen evolution rate of approximately 30% compared to that of the wild-type. The S₁ state Mn-XANES and Mn-EXAFS and S₂ state multiline EPR signals of purified D1-E333Q PSII core complexes closely resembled those of wild-type, aside from having lower amplitudes. The S(n+1)-minus-S(n) FTIR difference spectra showed only minor alterations to the carbonyl, amide, and carboxylate stretching regions. However, the mutation eliminated a negative peak at 3663 cm⁻¹ in the weakly H-bonding O-H stretching region of the S₂-minus-S₁ FTIR difference spectrum and caused an approximately 9 cm⁻¹ downshift of the negative feature in this region of the S₁-minus-S₀ FTIR difference spectrum. We conclude that fully functional Mn₄CaO₅ clusters assemble in the presence of the D1-E333Q mutation but that the mutation decreases the yield of assembled clusters and alters the H-bonding properties of one or more water molecules or hydroxide groups that are located on or near the Mn₄CaO₅ cluster and that either deprotonate or form stronger hydrogen bonds during the S₀ to S₁ and S₁ to S₂ transitions.
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis