Merck
Local insulin therapy affects fracture healing in a rat model.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2012-12-15)
Andrew G Park, David N Paglia, Loay Al-Zube, Jeremy Hreha, Swaroopa Vaidya, Eric Breitbart, Joseph Benevenia, J Patrick O'Connor, Sheldon S Lin
ABSTRACT
A significant number of lower extremity fractures result in mal-union necessitating effective treatments to restore ambulation. Prior research in diabetic animal fracture models demonstrated improved healing following local insulin application to the fracture site and indicated that local insulin therapy can aid bone regeneration, at least within an insulin-dependent diabetic animal model. This study tested whether local insulin therapy could accelerate femur fracture repair in normal, non-diabetic rats. High (20 units) and low (10 units) doses of insulin were delivered in a calcium sulfate carrier which provided sustained release of the exogenous insulin for 7 days after fracture. Histomorphometry, radiographic scoring, and torsional mechanical testing were used to measure fracture healing. The fracture calluses from rats treated with high-dose insulin had significantly more cartilage than untreated rats after 7 and 14 days of healing. After 4 weeks of healing, femurs from rats treated with low-dose insulin had significantly higher radiographic scores and mechanical strength (p < 0.05), compared to the no treatment control groups. The results of this study suggest that locally delivered insulin is a potential therapeutic agent for treating bone fractures. Further studies are necessary, such as large animal proof of concepts, prior to the clinical use of insulin for bone fracture treatment.
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%