Merck
  • Chemical induction of endogenous retrovirus particles from the vero cell line of African green monkeys.
Chemical induction of endogenous retrovirus particles from the vero cell line of African green monkeys.
Journal of virology (2011-05-06)
Hailun Ma, Yunkun Ma, Wenbin Ma, Dhanya K Williams, Teresa A Galvin, Arifa S Khan
ABSTRACT
Endogenous retroviral sequences are present in high copy numbers in the genomes of all species and may be expressed as RNAs; however, the majority are defective for virus production. Although virus has been isolated from various Old World monkey and New World monkey species, there has been no report of endogenous retroviruses produced from African green monkey (AGM) tissues or cell lines. We have recently developed a stepwise approach for evaluating the presence of latent viruses by chemical induction (Khan et al., Biologicals 37:196-201, 2009). Based upon this strategy, optimum conditions were determined for investigating the presence of inducible, endogenous retroviruses in the AGM-derived Vero cell line. Low-level reverse transcriptase activity was produced with 5-azacytidine (AzaC) and with 5'-iodo-2'-deoxyuridine (IUdR); none was detected with sodium butyrate. Nucleotide sequence analysis of PCR-amplified fragments from the gag, pol, and env regions of RNAs, prepared from ultracentrifuged pellets of filtered supernatants, indicated that endogenous retrovirus particles related to simian endogenous type D betaretrovirus (SERV) sequences and baboon endogenous virus type C gammaretrovirus (BaEV) sequences were induced by AzaC, whereas SERV sequences were also induced by IUdR. Additionally, sequence heterogeneity was seen in the RNAs of SERV- and BaEV-related particles. Infectivity analysis of drug-treated AGM Vero cells showed no virus replication in cell lines known to be susceptible to type D simian retroviruses (SRVs) and to BaEV. The results indicated that multiple, inducible endogenous retrovirus loci are present in the AGM genome that can encode noninfectious, viruslike particles.
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)
Roche
Taq DNA Polymerase, 1 U/μl, suitable for PCR, hotstart: no, dNTPs included: no
Sigma-Aldrich
5-Azacytidine, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)