Merck
  • Comparison of SSI with APCI as an interface of HPLC-mass spectrometry for analysis of a drug and its metabolites.
Comparison of SSI with APCI as an interface of HPLC-mass spectrometry for analysis of a drug and its metabolites.
Journal of the American Society for Mass Spectrometry (2002-03-23)
Tetsuya Arinobu, Hideki Hattori, Hiroshi Seno, Akira Ishii, Osamu Suzuki
ABSTRACT
Sonic spray ionization (SSI) was compared with atmospheric pressure chemical ionization (APCI) as an interface of high-performance liquid chromatography (HPLC)-mass spectrometry (MS) for sensitive analyses of a neuroleptic drug, haloperidol and its two metabolites, such as reduced haloperidol and 4-(4-chlorophenyl)-4-hydroxypiperidine (CPHP), in biological samples. For both SSI and APCI interfaces, HPLC-MS-MS gave higher sensitivity than HPLC-MS. The sensitivities by HPLC-SSI-MS-MS for haloperidol and reduced haloperidol were 100 and 30 times higher, respectively, than those by HPLC-APCI-MS-MS; no spectrum with recognizable peaks was obtained for CPHP with the APCI interface. Therefore, detection limits and regression equations were examined by the HPLC-SSI-MS-MS for human plasma and urine samples spiked with the above drug and its metabolites. Haloperidol, reduced haloperidol, and CPHP showed good linearity in the ranges of 5-800, 10-800, and 100-800 ng/mL, respectively, for both human plasma and urine; their detection limits were 2.5, 5, and 75 ng/mL, respectively, using a new polymer HPLC column which enabled direct application of biological samples.
Sigma-Aldrich
4-(4-Chlorophenyl)-4-hydroxypiperidine, 99%
Supelco
Haloperidol metabolite II, analytical standard