Merck
  • Hyponatremia-related liver steatofibrosis and impaired spermatogenesis: evidence from a mouse model of the syndrome of inappropriate antidiuresis.
Hyponatremia-related liver steatofibrosis and impaired spermatogenesis: evidence from a mouse model of the syndrome of inappropriate antidiuresis.
Journal of endocrinological investigation (2022-11-28)
G Marroncini, C Anceschi, L Naldi, B Fibbi, M Brogi, L Lanzilao, A Fanelli, M Maggi, A Peri
ABSTRACT
Hyponatremia is the most frequent electrolytic disorder in clinical practice. In addition to neurological symptoms, hyponatremia, even when mild/moderate and chronic, has been related to other manifestations, such as bone demineralization and increased risk of fractures. To better elucidate tissue alterations associated with reduced serum sodium concentration [Na+], we developed an in vivo model of hyponatremia secondary to the Syndrome of Inappropriate Antidiuresis. Hyponatremia was induced in Foxn1nu/nu mice by subcutaneous infusion of the vasopressin analog 1-deamino [8-D-arginine] vasopressin (dDAVP) for 14 days via osmotic mini-pumps. Mice in the control group were infused with isotonic saline solution. Serum [Na+] progressively decreased, with a nadir of 123.4 ± 2.3 mEq/L (mean ± SD, dDAVP 0.3 ng/h) and 111.6 ± 4.7 mEq/L (mean ± SD, dDAVP 0.5 ng/h). Evident signs of liver steatofibrosis were observed at histology in hyponatremic mice. Accordingly, the expression of proteins involved in lipid metabolism (SREBP-1, PPARα and PPARγ) and in myofibroblast formation (αSMA and CTGF) significantly increased. Furthermore, heme oxygenase 1 expression was up-regulated in Kupffer and hepatic stellate cells in the liver of hyponatremic mice. Testis alterations were also observed. In particular, the thickness of the seminiferous epithelium appeared reduced. The expression levels of PCNA and PTMA, which are involved in DNA replication and germ cells maturation, were markedly reduced in the testis of hyponatremic mice. Overall, these findings shed new light on the possible consequences of chronic hyponatremia and prompt a more thorough evaluation of hyponatremic patients.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-PTMA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HMOX1, affinity isolated antibody