Merck
  • Multi-omic profiling of primary mouse neutrophils predicts a pattern of sex and age-related functional regulation.
Multi-omic profiling of primary mouse neutrophils predicts a pattern of sex and age-related functional regulation.
Nature aging (2021-09-14)
Ryan J Lu, Shalina Taylor, Kévin Contrepois, Minhoo Kim, Juan I Bravo, Mathew Ellenberger, Nirmal K Sampathkumar, Bérénice A Benayoun
ABSTRACT
Neutrophils are the most abundant human white blood cell and constitute a first line of defense in the innate immune response. Neutrophils are short-lived cells, and thus the impact of organismal aging on neutrophil biology, especially as a function of biological sex, remains poorly understood. Here, we describe a multi-omic resource of mouse primary bone marrow neutrophils from young and old female and male mice, at the transcriptomic, metabolomic and lipidomic levels. We identify widespread regulation of neutrophil 'omics' landscapes with organismal aging and biological sex. In addition, we leverage our resource to predict functional differences, including changes in neutrophil responses to activation signals. To date, this dataset represents the largest multi-omics resource for neutrophils across sex and ages. This resource identifies neutrophil characteristics which could be targeted to improve immune responses as a function of sex and/or age.
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Ready Made solution, 1 mg/mL