Merck
  • Transforming growth factor-beta and liver injury in an arginine vasopressin-induced pregnant rat model.
Transforming growth factor-beta and liver injury in an arginine vasopressin-induced pregnant rat model.
Clinical and experimental reproductive medicine (2021-05-25)
Nalini Govender, Sapna Ramdin, Rebecca Reddy, Thajasvarie Naicker
ABSTRACT
Approximately 30% of preeclamptic pregnancies exhibit abnormal liver function tests. We assessed liver injury-associated enzyme levels and circulating transforming growth factor beta (TGF-β) levels in an arginine vasopressin (AVP)-induced pregnant Sprague-Dawley rat model. Pregnant and non-pregnant Sprague-Dawley rats (n=24) received AVP (150 ng/hr) subcutaneously via mini-osmotic pumps for 18 days. Blood pressure was measured, urine samples were collected, and all animals were euthanized via isoflurane. Blood was collected to measure circulating levels of TGF-β1-3 isomers and liver injury enzymes in pregnant AVP (PAVP), pregnant saline (PS), non-pregnant AVP (NAVP), and non-pregnant saline (NS) rats. The PAVP group showed significantly higher systolic and diastolic blood pressure than both saline-treated groups. The weight per pup was significantly lower in the AVP-treated group than in the saline group (p<0.05). Circulating TGF-β1-3 isomer levels were significantly higher in the PAVP rats than in the NS rats. However, similar TGF-β1 and TGF-β3 levels were noted in the PS and PAVP rats, while TGF-β2 levels were significantly higher in the PAVP rats. Circulating liver-type arginase-1 and 5'-nucleotidase levels were higher in the PAVP rats than in the saline group. This is the first study to demonstrate higher levels of TGF-β2, arginase, and 5'-nucleotidase activity in PAVP than in PS rats. AVP may cause vasoconstriction and increase peripheral resistance and blood pressure, thereby elevating TGF-β and inducing the preeclampsia-associated inflammatory response. Future studies should explore the mechanisms through which AVP dysregulates liver injury enzymes and TGF-β in pregnant rats.
Product Description
Millipore
MILLIPLEX® TGFß Magnetic Bead 3 Plex Kit - Immunology Multiplex Assay, for quantification of TGFß1, TGFß2 and TGFß3 in multiple species
Millipore
MILLIPLEX® Rat Liver Injury Panel - Toxicity Multiplex Assay