Merck
  • Training rhesus macaques to take daily oral antiretroviral therapy for preclinical evaluation of HIV prevention and treatment strategies.
Training rhesus macaques to take daily oral antiretroviral therapy for preclinical evaluation of HIV prevention and treatment strategies.
PloS one (2019-11-16)
Michele B Daly, April M Clayton, Susan Ruone, James Mitchell, Chuong Dinh, Angela Holder, Julian Jolly, J Gerardo García-Lerma, James L Weed
ABSTRACT
Macaque models of simian or simian/human immunodeficiency virus (SIV or SHIV) infection are critical for the evaluation of antiretroviral (ARV)-based HIV treatment and prevention strategies. However, modelling human oral ARV administration is logistically challenging and fraught by limited adherence. Here, we developed a protocol for administering daily oral doses of ARVs to macaques with a high rate of compliance. Parameters of positive reinforcement training (PRT), behavioral responses and optimal drug delivery foods were defined in 7 male rhesus macaques (Macaca mulatta). Animals were trained to sit in a specified cage location prior to receiving ARVs, emtricitabine (FTC) and tenofovir alafenamide (TAF), in a blended food mixture, which was followed immediately with a juice chaser. Consistency of daily oral adherence was evaluated in 4 trained macaques receiving clinically equivalent doses of FTC and TAF (20 and 1.5 mg/kg, respectively) in a short-term (1 month) and an extended (6 month) trial. Adherence was monitored using medication diaries and by quantifying intracellular FTC-triphosphate (FTC-TP) and tenofovir-diphosphate (TFV-DP) concentrations in peripheral mononuclear blood cells (PBMCs). Trained macaques quickly and consistently took daily oral ARVs for 1 month with an average 99.8% observed adherence. Intracellular concentrations of TFV-DP (median = 845.8 fmol/million cells [range, 620.8-1031.3]) and FTC-TP (median = 367.0 fmol/million cells [range, 289.5-413.5) in PBMCs were consistent with high adherence. Extended treatment with select subjects yielded similar observations for three months (99.5% adherence, 352/356 complete doses taken), although a sudden drop in adherence was observed after splenic biopsy surgery. We demonstrate that trained macaques reliably adhere to a daily oral ARV regimen, although unexpected adherence issues are possible. Our approach, using clinical doses of oral FTC and TAF daily, further refines macaque models of HIV treatment and prevention by mimicking the human route and timing of ARV administration.
Sigma-Aldrich
Emtricitabine, ≥98% (HPLC)
Pricing and availability is not currently available.