Merck
  • Detecting Secretory Proteins by Acoustic Droplet Ejection in Multiplexed High-Throughput Applications.
Detecting Secretory Proteins by Acoustic Droplet Ejection in Multiplexed High-Throughput Applications.
ACS chemical biology (2019-01-31)
Michael J Iannotti, Ryan MacArthur, Richard Jones, Dingyin Tao, Ilyas Singeç, Sam Michael, James Inglese
ABSTRACT
Nearly one-third of the encoded proteome is comprised of secretory proteins that enable communication between cells and organ systems, playing a ubiquitous role in human health and disease. High-throughput detection of secreted proteins would enhance efforts to identify therapies for secretion-related diseases. Using the Z mutant of alpha-1 antitrypsin as a human secretory model, we have developed 1536-well high-throughput screening assays that utilize acoustic droplet ejection to transfer nanoliter volumes of sample for protein quantification. Among them, the acoustic reverse phase protein array (acoustic RPPA) is a multiplexable, low-cost immunodetection technology for native, endogenously secreted proteins from physiologically relevant model systems like stem cells that is compatible with plate-based instrumentation. Parallel assay profiling with the LOPAC1280 chemical library validated performance and orthogonality between a secreted bioluminescent reporter and acoustic RPPA method by consistently identifying secretory modulators with comparable concentration response relationships. Here, we introduce a robust, multiplexed drug discovery platform coupling extracellular protein quantification by acoustic RPPA with intracellular and cytotoxicity analyses from single wells, demonstrating proof-of-principle applications for human induced pluripotent stem cell-derived hepatocytes.
Sigma-Aldrich
Anti-α1-Antitrypsin antibody produced in rabbit, IgG fraction of antiserum