- The high-affinity transporter BnPHT1;4 is involved in phosphorus acquisition and mobilization for facilitating seed germination and early seedling growth of Brassica napus.
The high-affinity transporter BnPHT1;4 is involved in phosphorus acquisition and mobilization for facilitating seed germination and early seedling growth of Brassica napus.
Seed germination and seedling establishment are two of the most critical phases in plant development. However, the molecular mechanisms underlying the effect of phosphorus on seed germination and post-germinated growth of oilseed rape are unclear so far. Here, we report the role of BnPHT1;4 in seed germination and early seedling development of Brassica napus. Our results show that BnPHT1;4 is preferentially expressed in cotyledons of early developing seedlings. Overexpression of BnPHT1;4 in oilseed rape promoted seed germination and seedling growth. Expression levels of the genes related to ABA and GA biosynthesis and signaling were significantly altered in BnPHT1;4 transgenic seedlings. Consequently, active GA level was up-regulated, whereas ABA content was down-regulated in BnPHT1;4 transgenic seedlings. Furthermore, exogenous GA could promote seed germination of wild type, while exogenous ABA could partially recover the advanced-germination phenotype of BnPHT1;4 transgenic seeds. Total phosphorus content in cotyledons of the transgenic seedlings was decreased more rapidly than that in wild type when Pi was supplied or deficient, and Pi contents in shoots and roots of the BnPHT1;4 transgenic plants were higher than those in wild type under high and low Pi conditions. Our data suggest that the high-affinity transporter BnPHT1;4 is involved in phosphorus acquisition and mobilization for facilitating seed germination and seedling growth of Brassica napus by modulating ABA and GA biosynthesis.