Merck
  • Chronic corticosterone administration down-regulates metabotropic glutamate receptor 5 protein expression in the rat hippocampus.
Chronic corticosterone administration down-regulates metabotropic glutamate receptor 5 protein expression in the rat hippocampus.
Neuroscience (2010-07-06)
A H Iyo, A M Feyissa, A Chandran, M C Austin, S Regunathan, B Karolewicz
ABSTRACT
Several lines of evidence suggest a dysfunctional glutamate system in major depressive disorder (MDD). Recently, we reported reduced levels of metabotropic glutamate receptor subtype 5 (mGluR5) in postmortem brains in MDD, however the neurobiological mechanisms that induce these abnormalities are unclear. In the present study, we examined the effect of chronic corticosterone (CORT) administration on the expression of mGluR5 protein and mRNA in the rat frontal cortex and hippocampus. Rats were injected with CORT (40 mg/kg s.c.) or vehicled once daily for 21 days. The expression of mGluR5 protein and mRNA was assessed by Western blotting and quantitative real-time PCR (qPCR). In addition, mGluR1 protein was measured in the same animals. The results revealed that while there was a significant reduction (-27%, P=0.0006) in mGluR5 protein expression in the hippocampus from CORT treated rats, mRNA levels were unchanged. Also unchanged were mGluR5 mRNA and protein levels in the frontal cortex and mGluR1 protein levels in both brain regions. Our findings provide the first evidence that chronic CORT exposure regulates the expression of mGluR5 and are in line with previous postmortem and imaging studies showing reduced mGluR5 in MDD. Our findings suggest that elevated levels of glucocorticoids may contribute to impairments in glutamate neurotransmission in MDD.
Sigma-Aldrich
Metabotropic Glutamate Receptor 5, control peptide for AB5675
Sigma-Aldrich
Anti-mGluR1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Metabotropic Glutamate Receptor 5 Antibody, pain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®