Merck
  • Learning induces the translin/trax RNase complex to express activin receptors for persistent memory.
Learning induces the translin/trax RNase complex to express activin receptors for persistent memory.
eLife (2017-09-21)
Alan Jung Park, Robbert Havekes, Xiuping Fu, Rolf Hansen, Jennifer C Tudor, Lucia Peixoto, Zhi Li, Yen-Ching Wu, Shane G Poplawski, Jay M Baraban, Ted Abel
ABSTRACT
Long-lasting forms of synaptic plasticity and memory require de novo protein synthesis. Yet, how learning triggers this process to form memory is unclear. Translin/trax is a candidate to drive this learning-induced memory mechanism by suppressing microRNA-mediated translational silencing at activated synapses. We find that mice lacking translin/trax display defects in synaptic tagging, which requires protein synthesis at activated synapses, and long-term memory. Hippocampal samples harvested from these mice following learning show increases in several disease-related microRNAs targeting the activin A receptor type 1C (ACVR1C), a component of the transforming growth factor-β receptor superfamily. Furthermore, the absence of translin/trax abolishes synaptic upregulation of ACVR1C protein after learning. Finally, synaptic tagging and long-term memory deficits in mice lacking translin/trax are mimicked by ACVR1C inhibition. Thus, we define a new memory mechanism by which learning reverses microRNA-mediated silencing of the novel plasticity protein ACVR1C via translin/trax.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ALK7 Antibody, Upstate®, from rabbit