
Valerenic acid

Synonym(s):
(2E)-3-[(4S,7R,7aR)-2,4,5,6,7,7a-Hexadydro-3,7-dimethyl- 1H-inden-4-yl]-2-methyl-2-propenoic acid
Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4