
TOYOPEARL GigaCap® S-650M Bulk Media

Synonym(s):
TOYOPEARL GigaCap® S-650 Cation Exchange Media
Products
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing