
Staphylococcus aureus subsp. aureus WDCM 00032 Vitroids™

Products
Product Category
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing