
Psoralen

Synonym(s):
6,7-Furanocoumarin, 7H-Furo[3,2-g]benzopyran-7-one, Ficusin, Furo[3,2-g]coumarin
Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing