Polydatin
Synonym(s):
Piceid, Reservatrol-3-β-mono-D-glucoside, Resveratrol 3-O-β-D-glucopyranoside, Polydatin
Empirical Formula (Hill Notation):
C20H22O8
CAS Number:
Molecular Weight:
390.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing