
Plastic additive 9

Synonym(s):
Didodecyl 3,3′-thiodipropionate, Thiodipropionic acid dilauryl ester
Linear Formula:
S[CH2CH2CO2(CH2)11CH3]2
CAS Number:
Molecular Weight:
514.84
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2