Skip to Content
Merck
vibrant-m

Pamidronate disodium

vibrant-m
Synonym(s):
Pamidronate disodium pentahydrate
Empirical Formula (Hill Notation):
C3H9NNa2O7P2 · 5H2O
CAS Number:
Molecular Weight:
369.11

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing