N-Methyl-D-aspartic acid
Synonym(s):
(R)-2-(Methylamino)succinic acid, NMDA
Empirical Formula (Hill Notation):
C5H9NO4
CAS Number:
Molecular Weight:
147.13
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing