
Mitoxantrone hydrochloride

Synonym(s):
Mitoxantrone dihydrochloride, 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione dihydrochloride
Empirical Formula (Hill Notation):
C22H28N4O6 · 2HCl
CAS Number:
Molecular Weight:
517.40
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2