
Methylprednisolone acetate for peak identification

Synonym(s):
11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-acetate
Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing