Skip to Content
Merck
Metergoline

Metergoline

Metergoline
Synonym(s):
N-CBZ-[(8β)-1,6-Dimethylergolin-8-yl]methylamine, [(8β)-1,6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester
Empirical Formula (Hill Notation):
C25H29N3O2
CAS Number:
Molecular Weight:
403.52
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing