Skip to Content
Merck
MCPA-2-ethylhexyl ester PESTANAL®, analytical standard

MCPA-2-ethylhexyl ester

MCPA-2-ethylhexyl ester PESTANAL®, analytical standard
Synonym(s):
2-Ethylhexyl (4-chloro-2-methylphenoxy)acetate
Empirical Formula (Hill Notation):
C17H25ClO3
CAS Number:
Molecular Weight:
312.83
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reaction Type
Reagent Type
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing