
Manganese(0) carbonyl

Synonym(s):
Decacarbonyl dimanganese(0), Dimanganese(0) decacarbonyl
Linear Formula:
Mn2(CO)10
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing