
L-Thyroxine (T4) solution

Synonym(s):
L-Thyroxine, 3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4
Linear Formula:
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
776.87
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
Optical Isomer
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing