
Levalbuterol Related Compound D

Synonym(s):
5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzaldehyde hemisulfate salt, 5-{2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl}-2-hydroxybenzaldehyde hemisulfate salt
Linear Formula:
(C13H19NO3)2 · H2SO4
CAS Number:
Molecular Weight:
572.67
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing