
KT203

Synonym(s):
4′-[1-[[2-(Phenylmethyl)-1-piperidinyl]carbonyl]-1H-1,2,3-triazol-4-yl]-[1,1′-biphenyl]-3-carboxylic acid
Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing