
Goserelin

Synonym(s):
Goserelin acetate, [D-Ser(tBu)6,Azagly10]-LHRH acetate salt
Empirical Formula (Hill Notation):
C59H84N18O14
CAS Number:
Molecular Weight:
1269.41
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing