
Ethyl 3,4,5-trihydroxybenzoate

Synonym(s):
Ethyl gallate
Linear Formula:
3,4,5-(OH)3C6H2COOC2H5
CAS Number:
Molecular Weight:
198.17
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing