Skip to Content
Merck
Eosin B Dye content 95 %

Eosin B

Eosin B Dye content 95 %
Synonym(s):
Acid Red 91
Empirical Formula (Hill Notation):
C20H8Br2N2O9
CAS Number:
Molecular Weight:
580.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing