
Dirithromycin

Synonym(s):
[9S(R)]-9-Deoxo-11-deoxy-9,11-[imino[2-(2-methoxy)ethylidene]oxy]erthromycin; LY-237216
Empirical Formula (Hill Notation):
C42H78N2O14
CAS Number:
Molecular Weight:
835.07
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing