
(+)-DIP-Chloride™

Synonym(s):
(+)-B-Chlorodiisopinocampheylborane, (+)-Ipc2BCl
Empirical Formula (Hill Notation):
C20H34BCl
CAS Number:
Molecular Weight:
320.75
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing