
Dasatinib

Empirical Formula (Hill Notation):
C22H26ClN7O2S
CAS Number:
Molecular Weight:
488.01
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-2 of 2
Compare
Product Number
Description
Pricing